missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC38A5 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
140.00 EUR - 399.00 EUR
Specifications
Antigen | SLC38A5 |
---|---|
Concentration | 200 μg/mL |
Applications | Western Blot, ELISA |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16865995
|
Proteintech
28102-1-AP-20UL |
20 μL |
140.00 EUR
20µL |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16855995
|
Proteintech
28102-1-AP-150UL |
150 μL |
399.00 EUR
150µL |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The SLC38A5 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human SLC38A5. This antibody recognizes human, mouse, rat antigen. The SLC38A5 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
SLC38A5 | |
Western Blot, ELISA | |
Unconjugated | |
Human, Rat, Mouse | |
IgG |
200 μg/mL | |
Polyclonal | |
Rabbit | |
92745 | |
Primary |