Learn More
SLC38A5 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 28102-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
The SLC38A5 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human SLC38A5. This antibody recognizes human, mouse, rat antigen. The SLC38A5 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
SLC38A5 | |
Polyclonal | |
Unconjugated | |
150 μL | |
92745 | |
IgG |
Western Blot, ELISA | |
200 μg/mL | |
Rabbit | |
Primary | |
Human, Rat, Mouse |