Learn More
SLC38A5 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 28102-1-AP-150UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16855995
  • 399.00 EUR / 150µL
Estimated Shipment: 04-06-2024
Description
The SLC38A5 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human SLC38A5. This antibody recognizes human, mouse, rat antigen. The SLC38A5 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
SLC38A5
Polyclonal
Unconjugated
150 μL
92745
IgG
Western Blot, ELISA
200 μg/mL
Rabbit
Primary
Human, Rat, Mouse