missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC35D3 Antibody, DyLight 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-81839G
This item is not returnable.
View return policy
Description
SLC35D3 Polyclonal antibody specifically detects SLC35D3 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
| SLC35D3 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, ELISA, Immunofluorescence | |
| DyLight 488 | |
| bA55K22.3, frc, fringe-like 1, FRCL1, Fringe connection-like protein 1, MGC102873, solute carrier family 35 member D3, solute carrier family 35, member D3 | |
| SLC35D3 antibody was raised against a 17 amino acid synthetic peptide near the carboxy terminus of human SLC35D3. The immunogen is located within the last 50 amino acids of SLC35D3. Amino Acid Squence: LVRGTRYMKKDYLIENE | |
| 0.1 mL | |
| Cancer, Endocrinology, Signal Transduction | |
| 340146 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction