Learn More
SKLB-23bb, MedChemExpress
Product Code. 16497648
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16497648 10 mg 10mg
16460118 5 mg 5mg
16447556 100 mg 100mg
16413667 50 mg 50mg
16411759 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 16497648 Supplier Medchem Express Supplier No. HY18947
MedChemExpress SKLB-23bb is a potent and selective inhibitor for HDAC6 with an IC50 of 17 nM and shows 25-fold and 200-fold selectivity relative to HDAC1 (IC50=422 nM) and HDAC8 (IC50=3398 nM), respectively.
SKLB-23bb is a potent and selective inhibitor for HDAC6 with an IC50 of 17 nM and shows 25-fold and 200-fold selectivity relative to HDAC1 (IC50=422 nM) and HDAC8 (IC50=3398 nM), respectively.
Specifications
Chemical Name or Material SKLB-23bb
CAS 1815580-06-3
Color Yellow
Purity Grade Notes Research
Molecular Formula C21H24N4O4
Quantity 10 mg
Solubility Information DMSO : 32 mg/mL (80.72 mM; Need ultrasonic and warming)
SMILES CN(C1=CC(OCCCC(NO)=O)=C(OC)C=C1)C2=NC(C)=NC3=CC=CC=C32
Molecular Weight (g/mol) 396.44
Formula Weight 396.44
Percent Purity 99.37%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death