missing translation for 'onlineSavingsMsg'
Learn More

SKLB-23bb, MedChemExpress

Product Code. 16411759
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16411759 1 mg 1mg
16497648 10 mg 10mg
16460118 5 mg 5mg
16447556 100 mg 100mg
16413667 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16411759 Supplier Medchem Express Supplier No. HY18947

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress SKLB-23bb is a potent and selective inhibitor for HDAC6 with an IC50 of 17 nM and shows 25-fold and 200-fold selectivity relative to HDAC1 (IC50=422 nM) and HDAC8 (IC50=3398 nM), respectively.

SKLB-23bb is a potent and selective inhibitor for HDAC6 with an IC50 of 17 nM and shows 25-fold and 200-fold selectivity relative to HDAC1 (IC50=422 nM) and HDAC8 (IC50=3398 nM), respectively.

Specifications

Chemical Name or Material SKLB-23bb
CAS 1815580-06-3
Color Yellow
Purity Grade Notes Research
Molecular Formula C21H24N4O4
Quantity 1 mg
Solubility Information DMSO : 32 mg/mL (80.72 mM; Need ultrasonic and warming)
SMILES CN(C1=CC(OCCCC(NO)=O)=C(OC)C=C1)C2=NC(C)=NC3=CC=CC=C32
Molecular Weight (g/mol) 396.44
Formula Weight 396.44
Percent Purity 99.37%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.