missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SKF 38393 hydrochloride, MedChemExpress
Click to view available options
Quantity:
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
200mg
500mg
50mg
Description
SKF 38393 hydrochloride is a selective agonist of the dopamine D1 receptor (D1DR) with an IC50 of 110 nM.
Specifications
Specifications
| Chemical Name or Material | SKF 38393 hydrochloride |
| CAS | 62717-42-4 |
| Color | Brown |
| Purity Grade Notes | Research |
| Molecular Formula | C16H18ClNO2 |
| Quantity | 50 mg |
| Synonym | (±)-SKF-38393 hydrochloride SKF-38393A |
| Solubility Information | DMSO : ≥ 34 mg/mL (116.53 mM) ∣H2O : 10 mg/mL (34.27 mM; Need ultrasonic) |
| SMILES | OC1=C(O)C=C2C(C3=CC=CC=C3)CNCCC2=C1.[H]Cl |
| Molecular Weight (g/mol) | 291.77 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction