Learn More
SKF 38393 hydrochloride, MedChemExpress
Product Code. 16475906
Encompass_Preferred
Click to view available options
Quantity:
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
200mg
500mg
50mg
Product Code. 16475906
Brand: Medchem Express HY12520A
MedChemExpress SKF 38393 hydrochloride is a selective agonist of the dopamine D1 receptor (D1DR) with an IC50 of 110 nM.
SKF 38393 hydrochloride is a selective agonist of the dopamine D1 receptor (D1DR) with an IC50 of 110 nM.
Specifications
Chemical Name or Material SKF 38393 hydrochloride
CAS 62717-42-4
Color Brown
Purity Grade Notes Research
Molecular Formula C16H18ClNO2
Quantity 100 mg
Synonym (±)-SKF-38393 hydrochloride SKF-38393A
Solubility Information DMSO : ≥ 34 mg/mL (116.53 mM) ∣H2O : 10 mg/mL (34.27 mM; Need ultrasonic)
SMILES OC1=C(O)C=C2C(C3=CC=CC=C3)CNCCC2=C1.[H]Cl
Molecular Weight (g/mol) 291.77
Formula Weight 291.77
Percent Purity 98.22%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease