Learn More
SIRT1 Monoclonal Antibody (OTI12H3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA809859
Encompass_Preferred
 View more versions of this product
Product Code. 17582464
  • 540.87 EUR / 100µL
Description
NAD-dependent protein deacetylase sirtuin-1 (SIRT1) links transcriptional regulation directly to intracellular energetics and participates in the coordination of several separated cellular functions such as cell cycle, repsonse to DNA damage, metobolism, apoptosis, and authopahy. SIRT1 can modulate chromatin function through deacetylation of histones and can promote alterations in the methylation of histone and DNA, leading to transcriptional repression. It is essential in skeletal muscle cell differentiation and in response to low nutrients mediates the inhibitory effect on skeletal myoblast differentiation which also involves 5′-AMP-activated protein kinase (AMPK) and nicotinamide phosphoribosyltransferase (NAMPT).
Specifications
Specifications
SIRT1
Monoclonal
Unconjugated
SIRT1
SIR2, SIR2L1, SIR2alpha
Mouse
Affinity Chromatography
RUO
23411
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
OTI12H3
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q96EB6
SIRT1
Human recombit protein fragment corresponding to amino acids 552-676 of human SIRT1 produced in E.coli.
100 μL
Primary
Human
Antibody
IgG2b
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions