missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SIRT1 Monoclonal Antibody (OTI12H3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA809859S
This item is not returnable.
View return policy
Description
SIRT1 Monoclonal antibody specifically detects SIRT1 in Human samples. It is validated for Western BlotSpecifications
| SIRT1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P37837 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 6888 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2b |
| Western Blot | |
| OTI12H3 | |
| Unconjugated | |
| TALDO1 | |
| 75SirT1; AA673258; hSIR2; hSIRT1; mSIR2a; NAD-dependent deacetylase sirtuin-1; NAD-dependent protein deacetylase sirtuin-1; OTTHUMP00000198111; OTTHUMP00000198112; Regulatory protein SIR2 homolog 1; RP11-57G10.3; SIR2; Sir2a; SIR2alpha; SIR2L1; sir2-like 1; SIR2-like protein 1; SIRT 1; SIRT1; SIRT-1; SirtT1 75 kDa fragment; sirtuin 1; sirtuin 1 ((silent mating type information regulation 2, homolog) 1; sirtuin type 1 | |
| Full length human recombinant protein of human TALDO1 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction