missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SIRPB1 Monoclonal Antibody (OTI3G8), TrueMAB™, OriGene
Description
Immunoglobulin-like cell surface receptor involved in the negative regulation of receptor tyrosine kinase-coupled signaling processes.
Specifications
Specifications
| Antigen | SIRPB1 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | OTI3G8 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide |
| Gene | SIRPB1 |
| Gene Accession No. | O00241 |
| Gene Alias | CD172b, SIRP-BETA-1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?