missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SIRPB1 Monoclonal Antibody (OTI3G8), TrueMAB™, OriGene
Description
SIRPB1 Monoclonal antibody specifically detects SIRPB1 in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | SIRPB1 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | OTI3G8 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide |
| Gene | DTWD1 |
| Gene Accession No. | Q8N5C7 |
| Gene Alias | 9930027N05Rik; CD172 antigen-like family member B; CD172b; CD172b antigen; DKFZp686A05192; signal regulatory protein beta 1; signal-regulatory protein beta 1; signal-regulatory protein beta 1A; signal-regulatory protein beta-1; Signal-regulatory protein beta-1 isoform 3; SIRP BETA 1; SIRP beta 1 cell surface protein; Sirpb; SIRPB1; Sirpb1a; SIRP-beta; SIRP-beta B; SIRP-BETA-1; SIRP-beta-1 isoform 3 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?