Learn More
Siremadlin (R Enantiomer), MedChemExpress
Product Code. 16470038
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
Unit Size:
10mg
1mg
50mg
5mg
Product Code. 16470038
Brand: Medchem Express HY18658A
MedChemExpress Siremadlin R Enantiomer (NVP-HDM201 R Enantiomer) is the R enantiomer of Siremadlin. Siremadlin is a potent and highly specific MDM-2/p53 inhibitor.
Siremadlin R Enantiomer (NVP-HDM201 R Enantiomer) is the R enantiomer of Siremadlin. Siremadlin is a potent and highly specific MDM-2/p53 inhibitor.
Specifications
Chemical Name or Material Siremadlin (R Enantiomer)
CAS 1448867-42-2
Color Off-White
Purity Grade Notes Research
Molecular Formula C26H24Cl2N6O4
Quantity 5 mg
Synonym NVP-HDM201 (R Enantiomer) HDM201 (R Enantiomer)
Solubility Information DMSO : ≥ 56.75 mg/mL (102.18 mM)
SMILES O=C1N(C2=CC(Cl)=CN(C)C2=O)[C@H](C3=CC=C(Cl)C=C3)C4=C1N=C(C5=CN=C(OC)N=C5OC)N4C(C)C
Molecular Weight (g/mol) 555.41
Formula Weight 555.41
Percent Purity 99.02%
Grade Research
Physical Form Solid