Learn More
Honeywell™ Single-Use Molded Facemask with Valve and Facial Seal, FFP3 NRD
Description
- Heat-molded shell: shape maintained even in moistened and heavy loading environments
- Soft and hypoallergenic full face seal with absorption properties, a tight sealing film and a unique design, highly adapted to the nasal area.
- Preformed and colored nosebridge to save time to perfectly adjust the mask and to instantly identify its protection level
- Large internal volume to easily communicate and breathe
- Shielded exhalation valve with an exclusive design for improved protection and breathing comfort
- Twin elastic straps, ultra flexible and strong, stapled outside the filtering area to avoid any risk of leakages
- Lightweight (18 g) for better wearer acceptance
- Free of latex, silicone, and PVC
Specifications
Specifications
| Design Type | Heat-molded shell |
| Size | Medium |
| For Use With (Application) | Automotive, building, construction, food, foundry, iron and steel, medical and pharmaceutical, mining, quarrrying, textile, wood industries |
| Includes | Double elastics, valve, facial seal |
| Latex Free | Yes |
| Weight (Metric) | 18 g |