Learn More
BINDER™ Single Measurement Temperature and Humidity Calibration Certificate
Shop All BINDER Products
Click to view available options
For Use With (Equipment):
KBF PRO Series
KBF Series
Unit Size:
Each
Description
Calibration measurement taken from the center of the chamber at 25°C/60% RH, or at specified test values.
Specifications
Specifications
| For Use With (Equipment) | KBF PRO Series |
| Duration | 1 Day |
| Product Type | Calibration Certificate |