Learn More
Thermo Scientific™ Single-cell sample holders for GENESYS™ and BioMate™ 160 Spectrophotometers
Shop All Thermo Scientific Products
Click to view available options
For Use With (Equipment):
4-position carousel base
GENESYS 140/150/180,BioMate160
GENESYS 30/40/50
Spectronic200,GENESYS 30/40/50
Type:
Beckman-style Cell Holder
Cylindrical Cell Holder
Disposable Microcell Holder
Hose Entry/Exit Kit
Indiv Adjustable Filter Holder
Long Path Cell Holder
Sample Holder for Film/Filter
Single Cell Sample Holder
Test Tube Cap
Test Tube and Vial Holder
Holds:
10 mm cuvette (1)
10 to 100 mm cuvette (1)
12 mm to 25 mm tubes and vials
20 to 100 mm cuvette (1)
5 - 100 mm cylindrical cells
5 to 100 mm cuvette (1)
5 to 100mm cylindrical cuvette
50 μL microcells, Uvettes
Beckman-style short microcells
Entry and exit hoses (incl)
Filter/lenses, 10 mm thick
Filter/lenses, 6 mm thick
Filters up to 10 mm thick
Single-cell cuvette, 10mm tube
Square 10 mm PL Cuvette
Tall tubes, vials (lid open)
Unit Size:
Each
Description
Make the most of every sample. Accessories for Thermo Scientific™ GENESYS™ UV-Vis spectrophotometers are designed to enable you to work smarter, not harder. We offer a wide variety of options to meet your measurment requirments. Cable-free, snap-in design allows for easy exchange of accessories.
We provide single-cell solutions for measuring different types of samples.
Get the most from your measurments with accessories for the GENESYS Vis/UV-Vis Spectrophotometer family. Simplify sampling with easy access, flexibility and choices to fit your varied testing needs.
Specifications
Specifications
| Description | Accessory Kit for Hose Entry/Exit |
| Type | Hose Entry/Exit Kit |
| For Use With (Application) | Thermostatting samples with flowing water. Flow-cell applications with an external pump or sipper. |
| For Use With (Equipment) | GENESYS 30/40/50 |
| Holds | Entry and exit hoses (incl) |
| Includes | Sample compartment and sample compartment lid, hoses |
| Required Accessories | Requires suitable cell holder such as thermostatted cell holder 335079-000 or standard 10 mm square cell holder 336028-000 |