Learn More
Silver Nitrate Solution R2, Ph. Eur. Grade, Reagecon™
BIOPHARMA
Product Code. 11434739 Shop All Reagecon Products
Click to view available options
Quantity:
1 L
Unit Size:
1 L
Product Code. 11434739
Brand: Reagecon 1078302
Specifications
Quantity 1 L
compliance-icons compliance-icons compliance-icons
Product Identifier
  • SILVER NITRATE SOLUTION R2
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P273-Avoid release to the environment.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P501-Dispose of contents/container to .
Supplemental information
  • MIXTURE LIST-Contains : silver nitrate