Learn More
Trajan Scientific and Medical™ SilTite™ Mini Union
Delivers an easy, leak-free installation for capillary columns without the use of any tools.
Brand:  Trajan Scientific and Medical™ 073554
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15238933
  • 125.00 EUR / Each
Estimated Shipment: 03-06-2024
to see stock.
Add to basket
Description
Description
  • Specifically designed for connecting capillary columns
  • Connects guard columns to capillary columns
  • Inert and precise glass lined bore
  • Low dead volume
Specifications
Specifications
Mini Union
Inert and Precise Glass Lined Bore
0.8 mm
Connecting Capillary Columns