missing translation for 'onlineSavingsMsg'
Learn More

Silthiofam 100 μg/mL in Ethyl acetate, Dr. Ehrenstorfer

Product Code. 30319425
Change view
Click to view available options
Quantity:
5 mL
Unit Size:
5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30319425 5 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30319425 Supplier Dr. Ehrenstorfer Supplier No. DREV16947000EA100

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Silthiofarm
CAS 175217-20-6
Analyte or Component Names Silthiofarm
Shipping Condition +20°C
Molecular Formula C13 H21 N O S Si
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
InChI Formula InChI=1S/C13H21NOSSi/c1-7-8-14-12(15)11-9(2)10(3)16-13(11)17(4,5)6/h7H,1,8H2,2-6H3,(H,14,15)
SMILES Cc1sc(c(C(=O)NCC=C)c1C)[Si](C)(C)C
IUPAC Name 4,5-dimethyl-N-prop-2-enyl-2-trimethylsilylthiophene-3-carboxamide
Solvent or Matrix Ethyl Acetate
Molecular Weight (g/mol) 267.46
Quantity 5 mL
Formula Weight 267.1113 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.