Learn More
Deutsch Neumann™ Silicone Rubber Stoppers
Product Code. p-8007994
Click to view available options
Diameter (Metric) Bottom:
18 mm
50.5 mm
56 mm
60 mm
64.5 mm
71 mm
79 mm
94 mm
Diameter (Metric) Top:
24 mm
59.5 mm
65 mm
70 mm
75.5 mm
83 mm
92 mm
107 mm
Unit Size:
Each
Product Code. 10717781
Brand: Deutsch Neumann™ 1010518
Only null left
Add to basket
Add to basket
For safe and secure closure of laboratory apparatus. Deutsch Neumann™ Silicone Rubber Stoppers are translucent in color.
  • Acc. to DIN 12 871
  • Durometer 50 Shore A
  • Available in a wide range of sizes
Specifications
Quantity 1/Ea.
Temperature Range (Metric) 200°C
Color Transparent
Diameter (Metric) Bottom 18 mm
Diameter (Metric) Top 24 mm
Material Silicone
Product Type Silicone Stopper