Learn More
Macherey-Nagel™ Silicone Septa
Product Code. 10505941 Shop All Macherey-Nagel Products
Click to view available options
Diameter (Metric) Outer:
9 mm
12 mm
13 mm
Unit Size:
Pack of 50
Product Code. 10505941
Brand: Macherey-Nagel™ 702602
Only null left
Specifications
Type Septum
Diameter (Metric) Outer 9 mm
Hardness 45° Shore A
Temperature (Metric) Max 250°C
Color Clear
Material Silicone
Quantity 50 Pack