Learn More
Lenz Laborglasintrumente Silicone Sealing Rings
Product Code. p-7222603
Click to view available options
Bore Size:
10 mm
12 mm
14 mm
16 mm
18 mm
32 mm
Unit Size:
Pack of 10
Product Code. 11536894
Brand: Lenz Laborglasintrumente 01333216
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • For screw caps with bore
  • Material: silicone rubber (VMQ)
  • With bonded PTFE face
Specifications
Bore Size 16 mm
For Use With (Equipment) For thread 32 GL
Diameter (Metric) Outside 29 mm
Material Silicone