Learn More
Lenz Laborglasintrumente Silicone Sealing Rings
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand:  Lenz Laborglasintrumente 01333210
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11506894
  • 129.32 EUR / Pack of 10
Estimated Shipment: 25-06-2024
Description
  • For screw caps with bore
  • Material: silicone rubber (VMQ)
  • With bonded PTFE face
Specifications
Specifications
10 mm
Silicone
29 mm
For thread 32 GL