Learn More
Fisherbrand™ Silicone Pump Tubing
Shop All Fisherbrand Products
Click to view available options
Diameter (Metric) Inner:
0.5 mm
0.8 mm
1.6 mm
3.2 mm
4.8 mm
6.4 mm
8 mm
9.6 mm
Thickness (Metric):
1.6 mm
2.4 mm
Unit Size:
15m
Specifications
Specifications
| Certifications/Compliance | Class 6 |
| Autoclavable | Autoclavable |
| Diameter (Metric) Inner | 4.8 mm |
| Material | Silicone |
| Overall Length (Metric) | 1500 cm |
| Thickness (Metric) | 2.4 mm |
| For Use With | Peristaltic Pump |
| Packaging Quantity | 1 Each |
| Color | Translucent |
| Product Type | Pump Tubing |