Learn More
Ebro Electronic™ Extension Cable for Thermocouple Type K
GREENER_CHOICE
Product Code. 10269659
Click to view available options
Length (Metric) Cable:
1 m
2.5 m
Unit Size:
Each
Product Code. 10269659
Brand: Ebro Electronic™ 13412626
Only null left
Add to basket
Add to basket
Silcone extension cable for thermocouple, Type K
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance RoHS Directive 2011/65/EU
Disposable Single-use
For Use With (Application) Extension cable for probes with lemosa connector
Height (Metric) 30 mm
Length (Metric) 250 mm
Length (Metric) Cable 1 m
Material Silicone
Model AN 140
Weight (Metric) 0.12 kg
Product Type Extension Cable
For Use With (Equipment) TFN 520
Width (Metric) 100 mm