missing translation for 'onlineSavingsMsg'
Learn More

Silicon Standard: Si @ 1000 μg/g in Hydrocarbon Oil, VHG™

Product Code. 17920876
Change view
Click to view available options
Quantity:
50 g
200 g
400 g
Unit Size:
200g
400g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17920876 200 g 200g
17990526 400 g 400g
17983766 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17920876 Supplier VHG Labs Supplier No. VHGOSI1000200G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

Silicon Standard: Si @ 1000 μg/g in Hydrocarbon Oil
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Silicon
CAS 7440-21-3
Methods ICP/XRF/RDE
Analyte or Component Names Silicon
Shipping Condition Room Temperature
Molecular Formula Si
Synonym Silicon
InChI Formula InChI=1S/Si
SMILES [Si]
IUPAC Name silicon
Solvent or Matrix 75 cSt Mineral Oil
Molecular Weight (g/mol) 28.086
Quantity 200 g
Formula Weight 27.977 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.