Learn More
Silicon AAS Standard, 1000 mg/L Si in water, Honeywell Fluka™
Product Code. 16644384 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
Product Code. 16644384
Brand: Honeywell Fluka E4068500ML
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Honeywell Fluka™ analytical standards offer a growing range of high quality inorganic and organic reference materials for the European market produced according to ISO 9001. The products are suitable for a wide range of industries and applications and come complete with a detailed Certificate of Analysis (CoA) in every delivery, and are eligible for free expert technical support ensuring you have the information you need to quickly start your analysis and pass an audit.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Silicon AAS Standard
Density 1 g/cm3
Quantity 500 mL
Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado