missing translation for 'onlineSavingsMsg'
Learn More

Palintest™ Silica Test Kit

Product Code. 11993647
Click to view available options
Quantity:
50 Pack
Unit Size:
Pack of 50
This item is not returnable. View return policy

Product Code. 11993647

Brand: Palintest™ PM181

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

No. of Tests 50
Quantity 50 Pack
Range 0 to 4mg/L
Sufficient For 50 Tests
Type Silica Test Kit
compliance-icons compliance-icons
Product Identifier
  • TEST KIT/PHOTOM SILICA 0-4PPM
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Reproductive toxicity Category 1B
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H360FD-May damage fertility. May damage the unborn child.
Precautionary Statement
  • P264-Wash contaminated skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
  • P405-Store locked up.
Supplemental information
  • MIXTURE LIST-Contains : boric acid, citric acid, sulphamic acid,ammonium molybdate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.