missing translation for 'onlineSavingsMsg'
Learn More

Silica Standard: SiO2 @ 100 μg/mL in H2O, VHG™

Product Code. 17988094
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17988094 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17988094 Supplier VHG Labs Supplier No. VHGISIO2100500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

Silica Standard: SiO2 @ 100 μg/mL in H2O
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Silicon dioxide
CAS 7631-86-9
Methods ICP/ICP-MS/Wet Chemistry
Analyte or Component Names Silicon dioxide
Shipping Condition Room Temperature
Molecular Formula O2 Si
Synonym Silica, Silica colloidal anhydrous, Silicium dioxide
InChI Formula InChI=1S/O2Si/c1-3-2
SMILES O=[Si]=O
IUPAC Name Silicon dioxide
Solvent or Matrix Water
Molecular Weight (g/mol) 60.0843
Quantity 500 mL
Formula Weight 59.9668 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.