missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SIK1/Snf1lk Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
168.00 EUR
Specifications
| Antigen | SIK1/Snf1lk |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunohistochemistry 5 μg/mL, Immunohistochemistry-Paraffin 5 μg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
Description
SIK1/Snf1lk Polyclonal specifically detects SIK1/Snf1lk in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| SIK1/Snf1lk | |
| Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunohistochemistry 5 μg/mL, Immunohistochemistry-Paraffin 5 μg/mL | |
| Polyclonal | |
| Rabbit | |
| Protein Kinase | |
| PBS with 0.02% Sodium Azide | |
| EC 2.7.11, EC 2.7.11.1, msk, myocardial SNF1-like kinase, salt-inducible kinase 1, Salt-inducible protein kinase 1, serine/threonine protein kinase, serine/threonine-protein kinase SIK1, Serine/threonine-protein kinase SNF1-like kinase 1, Serine/threonine-protein kinase SNF1LK, SIK, SIK-1, SNF1-like kinase, SNF1LKMSK | |
| SIK1 | |
| IgG | |
| Affinity Purified | |
| Two isoforms of SIK1 are known to exist; this antibody will detect both isoforms. SIK1 antibody is predicted to not cross-react with other SIK protein family members. |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human | |
| NP_775490 | |
| 150094 | |
| Antibody was raised against an 18 amino acid synthetic peptide near the center of human SIK1. The immunogen is located within amino acids 340 - 390 of SIK1. Amino Acid Squence: ARPGPARQPRPRSSDLS | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title