Learn More
Epredia™ Signature Series™ Clear-Rite 3™
Shop All Epredia Products
Click to view available options
Quantity:
4 x 1 gal.
Packungsgröße:
15.14 L
Beschreibung
Includes
(4) 1-gallon bottles
- Very low odor; Benzene-free
- Use in all tissue processors and automatic stainers
- Does not produce hard or brittle specimens, even after prolonged exposure
Spezifikation
Spezifikation
| Product Type | Clearing Agent |
| Quantity | 4 x 1 gal. |
Sicherheit und Handhabung
Signalwort
- Danger
- Flammable liquid Category 2
- Germ cell mutagenicity Category 1B
- Acute toxicity Category 1
- Specific target organ toxicity after repeated exposure Category 2
- H225-Highly flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H340-May cause genetic defects.
- H350-May cause cancer.
- H371-May cause damage to organs.
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P233-Keep container tightly closed.
- P240-Ground/bond container and receiving equipment.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P242-Use only non-sparking tools.
- P243-Take precautionary measures against static discharge.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P331-Do NOT induce vomiting.
- P370+P378-In case of fire: Use CO2, dry chemical, or foam to extinguish.
- P403+P235-Store in a well-ventilated place. Keep cool.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : proprietary mixtureof aliphatic hydrocarbons
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur