Learn More
Bohlender™ SICCO™ Glove Box Duo 2 with Transfer Chamber
Product Code. 17167674
Change view
Click to view available options
Material:
Acrylic Glass
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material unitSize
17167674 Acrylic Glass Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17167674 Supplier Bohlender™ Supplier No. V171008
Glove Box Duo 2 with Transfer Chamber with panels made of acrylic glass and base panel made of polyethylene.
  • Aluminum frame with panels made of acrylic glass, base panel made of polyethylene
  • Front and back side with one door each, two pairs of opposite glove ports each with one pair of natural rubber gloves size 9
  • Two cable lead-ins (left bottom side)
  • Transfer chamber (right) with two doors, external door with latch to prevent accidental opening
  • Tray extends into the workspace with opening of the internal door for easy removal of inserted products and is only operable from one workstation side (front right-hand)
  • Work space and transfer chamber include two hose connectors for gas-filling each and appropriate tubing (2 x 5 m tubing for gas-filling made of PVC and 2 x 5 m aeration tubing made of PE)
  • Robust and light-weight aluminum construction
  • Superb all-round visibility
  • Non-slip rubber feet
  • Cable lead-ins for electric devices
  • Large front opening
  • Connectors for gas-filling and aeration including appropriate tubing
  • Transfer chamber for inserting further products
Specifications
Material Acrylic Glass
Depth (English) 35 in.
Depth (English) Interior 33 in.
Depth (Metric) 890 mm
Depth (Metric) Interior 845 mm
Height (English) 28 in.
Height (English) Interior 26 in.
Height (Metric) 710 mm
Height (Metric) Interior 665 mm
Volume (Metric) 475 L
Weight (English) 66 lb.
Weight (Metric) 30 kg
Width (English) 47 in.
Width (English) Interior 33 in.
Width (Metric) 1200 mm
Width (Metric) Interior 850 mm