Learn More
SIBA, MedChemExpress
Description
SIBA (5'-Isobutylthioadenosine), a synthetic analogue of SAH (HY-19528), acts as an inhibitor of S-adenosylmethionine-mediated transmethylation. SIBA can interfere with a variety of enzymatic activities in vitro, such as SAH hydrolase, methylthioadenosine phosphorylase and cAMP phosphodiesterase. SIBA reversibly blocks the multiplication of herpes simplex type 1 virus (HSV).
Specifications
Specifications
| Chemical Name or Material | SIBA |
| CAS | 35899-54-8 |
| Color | Light Gray |
| Purity Grade Notes | Research |
| Molecular Formula | C14H21N5O3S |
| Quantity | 5 mg |
| Synonym | 5'-Isobutylthioadenosine 5'-Deoxy-5'-isobutylthioadenosine |
| Solubility Information | DMSO : ≥ 100 mg/mL (294.63 mM) |
| SMILES | O[C@@H]([C@H]([C@H](N1C=NC2=C1N=CN=C2N)O3)O)[C@H]3CSCC(C)C |
| Molecular Weight (g/mol) | 339.41 |
| Show More |