missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ Shrimp Alkaline Phosphatase (SAP)
Click to view available options
Quantity:
Each
Unit Size:
Each
Description
- 100% heat-inactivated in 5 min at 65°C
- Significantly improved storage stability at lower temperatures
- Very high specific activity
- Removes 5'-phosphates from DNA, RNA, dNTPs, and proteins
- Purified from a recombinant source
- May be added directly to restriction enzyme digests
- No vector purification necessary
- Requires no supplemental zinc or other additives for activity
- Works direct in many different buffers
- Easy treatment of unincorporated dNTPs in PCR products prior to DNA sequencing or SNP analysis
Specifications
Specifications
| Content And Storage | 37°C |
| Description | Shrimp Alkaline Phosphatase, 1 unit/μL Concentration, Tested for Contaminating Endonucleases, Exonucleases, and Ribonucleases |
| Detection Method | Tested for contaminating endonucleases, exonucleases, and ribonucleases |
| Final Product Type | Shrimp Alkaline Phosphatase |
| Product Type | Shrimp Alkaline Phosphatase |
| Quantity | Each |
| Purity | 0 |
For research use only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction