Learn More
Dräger™ Short-Term Detector Tubes - De Tube Hydrogen Sulfide
Product Code. 10388421
Click to view available options
Components Measured:
Hydrogen Sulfide
Hydrogen Sulfide 0.2%/a
Hydrogen Sulfide 0.2/b
Hydrogen Sulfide 2%/A
Hydrogen Sulfide 2/a
Hydrogen Sulfide 2/b
Hydrogen Sulfide, Sulfur Dioxide 0.2%/A
Measurement Range:
0.2 to 5ppm
0.2 to 6ppm
0.2 to 7 vol.%
2 to 200ppm
2 to 40 vol.%
2 to 60ppm
Unit Size:
Each
Pack of 10
Product Code. 10388421
Brand: Dräger™ CH28201
For gas detection
Note: Extension of measuring range listed is possible for some detector tubes. Call Fisher Customer Service for more information.
Specifications
Components Measured Hydrogen Sulfide, Sulfur Dioxide 0.2%/A
Humidity Range 40mg/L
Measurement Range 0.2 to 7 vol.%
No. of Strokes 1+2
Quantity 10
Response Time 2 min.
Temperature Range (English) 32°F to 104°F
Temperature Range (Metric) 0°C to 40°C