Learn More
Emma Safety Footwear Shoe Laces
Product Code. 10092154
Click to view available options
Color:
Black, Gray
White
Length (Metric):
95 cm
120 cm
Unit Size:
1 pair
Product Code. 10092154
Brand: Emma Safety Footwear MM00006100
Only null left
Add to basket
Add to basket
Available in White or black/gray colors.
Available in White or black/gray colors.
Specifications
Color White
Length (Metric) 95 cm
Product Type Shoe laces