Learn More
Emma Safety Footwear Shoe Laces
Click to view available options
Color:
Black, Gray
White
Length (Metric):
95 cm
120 cm
Unit Size:
1 pair
Description
Available in White or black/gray colors.
Specifications
Specifications
| Color | Black, Gray |
| Length (Metric) | 95 cm |
| Product Type | Shoe Laces |