Learn More
Shikonin, MedChemExpress
Product Code. 16483906
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16483906 25 mg 25mg
16474897 50 mg 50mg
16459118 10 mg 10mg
16419146 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16483906 Supplier Medchem Express Supplier No. HYN0822
MedChemExpress Shikonin is a major component of a Chinese herbal medicine named zicao. Shikonin is a potent TMEM16A chloride channel inhibitor with an IC50 of 6.5 μM. Shikonin is a specific pyruvate kinase M2 (PKM2) inhibitor and can also inhibit TNF-α and NF-κB pathway. Shikonin decreases exosome secretion through the inhibition of glycolysis. Shikonin inhibits AIM2 inflammasome activation.
Shikonin is a major component of a Chinese herbal medicine named zicao. Shikonin is a potent TMEM16A chloride channel inhibitor with an IC50 of 6.5 μM. Shikonin is a specific pyruvate kinase M2 (PKM2) inhibitor and can also inhibit TNF-α and NF-κB pathway. Shikonin decreases exosome secretion through the inhibition of glycolysis. Shikonin inhibits AIM2 inflammasome activation.
Specifications
Chemical Name or Material Shikonin
CAS 517-89-5
Color Dark Brown
Purity Grade Notes Research
Molecular Formula C16H16O5
Quantity 25 mg
Synonym C.I. 75535 Isoarnebin 4
Solubility Information DMSO : 25 mg/mL (86.72 mM; ultrasonic and warming and heat to 60°C)
SMILES O=C1C([C@H](O)C/C=C(C)/C)=CC(C2=C1C(O)=CC=C2O)=O
Molecular Weight (g/mol) 288.3
Formula Weight 288.3
Percent Purity 98.09%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease