Learn More
Thermo Scientific™ Shelves for Heratherm™ Ovens and Incubators
Shop All Thermo Scientific Products
Click to view available options
Depth (English):
11.4 in.
12.9 in.
14.8 in.
19.8 in.
20.2 in.
21.06 in.
Width (English):
11.4 in.
12.9 in.
14.8 in.
19.8 in.
20.2 in.
21.06 in.
38.74 in.
For Use With (Equipment):
Adv Prot OMH400/Gen Inc IGS400, 400l
Adv Prot OMH750/Gen Inc IGS750, 750l
Adv Prot/Gen Incubator 400l
Adv Prot/Gen Incubator 750l
Heratherm Adv Prot Incubator 100l
Heratherm Adv Prot Incubator 180l
Heratherm Adv Prot Oven 750I 400V
Heratherm Adv Protocol Incubator 60l
Heratherm Compact Incubator
Heratherm General Incubator 100L
Heratherm General Incubator 180l
Heratherm General Incubator 60l
Length (English):
12.9 in.
17.2 in.
20.7 in.
Unit Size:
Each
Description
Perforated and wire mesh shelves
Specifications
Specifications
| For Use With (Equipment) | Heratherm Adv Protocol Incubator 60l |
| Type | Shelf |
| Includes | (2) Shelf Supports |
| Material | Stainless Steel |
| No. of Shelves | 1 |
| Length (English) | 12.9 in. |
| Width (English) | 12.9 in. |
| Width (Metric) | 32.7 cm |
| Depth (English) | 12.9 in. |
| Depth (Metric) | 32.7 cm |