Learn More
BOLA Sheets
Product Code. p-200110021
Click to view available options
Width (Metric):
150 mm
300 mm
600 mm
Thickness:
0.025 mm
0.05 mm
0.25 mm
0.5 mm
0.75 mm
1 mm
1.5 mm
Unit Size:
Each
Product Code. 10470342
Brand: BOLA S180310
Only null left
Add to basket
Add to basket
Sheets, Non-absorbant
Delivered in rolls with a length of 1000 mm.
Specifications
Absorbancy Non-absorbent
Width (Metric) 300 mm
Thickness 0.75 mm