Learn More
Sheep anti-Rabbit IgG whole molecule, HRP, Polyclonal, Secondary Antibody, Abnova™
Brand: Abnova PAB10840.2mg
This item is not returnable.
View return policy
Specifications
| Sheep anti-Rabbit IgG whole molecule | |
| Polyclonal | |
| Sheep anti-rabbit IgG recognizes rabbit IgG whole molecule. This secondary antibody was purified using antigen affinity chromatography. The antibody is conjugated with Peroxidase. | |
| Lyophilized from 20mM potassium phosphate, 150mM NaCl, pH 7.2 (0.01% (w/v) gentamicin sulfate, 10mg/mL BSA (Sodium Azide, Immunoglobulin and Protease free)) | |
| Rabbit IgG whole molecule | |
| 2 mg | |
| Rabbit | |
| Lyophilized |
| Dot Blot, ELISA, Immunohistochemistry, Western Blot | |
| HRP | |
| The optimal working dilution should be determined by the end user. | |
| Sheep | |
| This product was prepared from monospecific antiserum by immunoaffinity chromatography, followed by solid phase adsorption(s) to remove any unwanted reactivities. | |
| Secondary | |
| Store at 4°C on dry atmosphere. After reconstitution with 1 mL of deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction