Learn More
Hospidex™ Sharpsafe™ Polypropylene Biohazard Container
Click to view available options
Capacity (Metric):
1 L
2 L
Unit Size:
Pack of 100
Pack of 50
Description
- Yellow
- Polypropylene
- NFX 30-505 certification
Specifications
Specifications
| Capacity (Metric) | 2 L |
| Certifications/Compliance | NFX30-500 |
| Height (Metric) | 168 mm |
| Length (Metric) | 196 mm |
| Autoclavable | Autoclavable |
| For Use With (Application) | For collection of sharps waste treatment activities, pungent, sharp and / or pointed |
| Material | Polypropylene |
| Product Type | Biohazard Container |