Learn More
Epredia™ Shandon™ Multi-Spot Slides
Shop All Epredia Products
Click to view available options
Description:
6mm Wells
9mm Wells
Unit Size:
Pack of 100
Specifications
Specifications
| Length (Metric) | 76 mm |
| Width (Metric) | 26 mm |
| Material | Glass |
| Type | Multispot Slide |
| Color | Plain |
| Description | 9mm Wells |
| Diameter (Metric) | 9 mm Wells |
| Height (Metric) | 9 mm |