Learn More
Epredia™ Glyo-Fixx™
Product Code. 12500067 Shop All Epredia Products
Click to view available options
Quantity:
1 gal.
125 x 60mL
5 gal.
125 x 120mL
Product Type:
Fixative
Unit Size:
3.8 L
Each
Pack of 125
Product Code. 12500067
Brand: Epredia™ 9990924
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Obtain superior results to neutral buffered formalin with Glyo-Fixx™, a noncarcinogenic, nonsensitizing, and environmentally friendly substitute.
  • Produces distinct and crisp cellular morphology
  • Fixes most antigens without impairing immunoreactivity
  • Allows special stain dyes, including silver procedures, to be sharp without background staining
  • Presents fine detail in specimen structure
TRUSTED_SUSTAINABILITY
Specifications
Product Type Fixative
Quantity 125 x 60mL