missing translation for 'onlineSavingsMsg'
Learn More

SH5-07, MedChemExpress

Product Code. 16464346
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16464346 100 mg 100mg
16499606 50 mg 50mg
16433048 10 mg 10mg
16410669 1 mg 1mg
16405787 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16464346 Supplier Medchem Express Supplier No. HY100494

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress SH5-07 is a hydroxamic acid based Stat3 inhibitor with an IC50 of 3.9 μM in in vitro assay.

SH5-07 is a hydroxamic acid based Stat3 inhibitor with an IC50 of 3.9 μM in in vitro assay.

Specifications

Chemical Name or Material SH5-07
CAS 1456632-41-9
Color White
Purity Grade Notes Research
Molecular Formula C29H28F5N3O5S
Quantity 100 mg
Solubility Information DMSO : 50 mg/mL (79.92 mM; Need ultrasonic)
SMILES O=C(NO)C1=CC=C(N(CC2=CC=C(C3CCCCC3)C=C2)C(CN(C)S(=O)(C4=C(F)C(F)=C(F)C(F)=C4F)=O)=O)C=C1
Molecular Weight (g/mol) 625.61
Formula Weight 625.61
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.