missing translation for 'onlineSavingsMsg'
Learn More

SH3D19 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Product Code. 16891085 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16891085 20 μL 20µL
16881085 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16891085 Supplier Proteintech Supplier No. 214991AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

May play a role in regulating A disintegrin and metalloproteases (ADAMs) in the signaling of EGFR-ligand shedding. May be involved in suppression of Ras-induced cellular transformation and Ras-mediated activation of ELK1. Plays a role in the regulation of cell morphology and cytoskeletal organization.

Specifications

Antigen SH3D19
Applications Immunofluorescence, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene SH3D19
Gene Accession No. Q5HYK7, Q91X43
Gene Alias ADAM binding protein Eve 1, EBP, EEN binding protein, EVE1, Kryn, SH3 domain containing 19, SH3D19, SH3P19
Gene Symbols SH3D19
Host Species Rabbit
Immunogen SH3D19 Fusion Protein Ag15915
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 152503, 27059, 295171
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.