missing translation for 'onlineSavingsMsg'
Learn More

SGX-523, MedChemExpress

Product Code. 16414928
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
2mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16414928

Brand: Medchem Express HY12019

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress SGX523 is a exquisitely selective and ATP-competitive MET inhibitor. SGX523 potently inhibits MET with an IC50 of 4 nM and is >1,000-fold selective versus other protein kinases. Antitumor activity.

SGX523 is a exquisitely selective and ATP-competitive MET inhibitor. SGX523 potently inhibits MET with an IC50 of 4 nM and is >1,000-fold selective versus other protein kinases. Antitumor activity.

Specifications

Chemical Name or Material SGX-523
Physical Form Solid
CAS 1022150-57-7
Color Light Yellow
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C18H13N7S
For Use With (Application) Cancer-Kinase/protease
Solubility Information DMSO : 30 mg/mL (83.47 mM; Need ultrasonic)
SMILES CN1N=CC(C2=NN3C(SC4=CC5=C(N=CC=C5)C=C4)=NN=C3C=C2)=C1
Molecular Weight (g/mol) 359.41
Formula Weight 359.41
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.