Learn More
Trajan™ Thermo Scientific AS3000, AS2000 and AS200/800GC Autosampler Syringe
Product Code. 15268463
Click to view available options
Quantity:
1/Pk.
6/Pk.
Unit Size:
Each
Product Code. 15268463
Brand: Trajan™ 0029861
Fixed needle autosampler syringes. For reduced non specific sample interaction and increased analyte recovery
  • High quality solutions for liquid handling
  • Accurate and reproducible injections
  • Specifically designed for Thermo Scientific instruments
Specifications
Needle Gauge 26 G
Diameter (Metric) Outer 0.47 mm
Product Type AS3000, AS2000 and AS200/800GC Autosampler Syringe
Quantity 6/Pk.
Volume (Metric) 10 μL
Length (Metric) Needle 50 mm