Learn More
SGE™ Series II Standard Fixed Needle Syringes
Product Code. 10559072
Click to view available options
Volume (Metric):
5 μL
10 μL
25 μL
50 μL
100 μL
250 μL
500 μL
Needle Gauge:
25 G
26 G
Includes:
Plunger protection
Standard plunger
Quantity:
10/Pk.
6 Pk.
Ea.
Unit Size:
Each
Pack of 10
Pack of 6
Product Code. 10559072
Brand: SGE™ 006000
Standard syringes with anti-roll flange and needles that are aligned with zero mark on barrel to reduce dead volume, improve accuracy and precision to <±1%
  • Stainless-steel plungers are hand fitted, noninterchangeable, nonreplaceable
  • Syringes with 5 and 10μL capacities have barrel O.D. of 6.5mm, scale length of 54.1mm and include a 50mm long 26s gauge needle
  • 25 to 500μL capacity syringes have 8mm O.D. barrel, 60mm scale length and 50mm long, 25 gauge needle Fixed-needle models have needle glued into barrel with solvent-resistant epoxy glue Removable-needle models feature a PTFE seal, a spring and nut for leak-resistant operation
Specifications
Accuracy ±1%
Material Borosilicate Glass (Barrel)
Needle Gauge 25 G
Diameter (English) Outer Needle 0.01 in.
Diameter (English) Outer 0.31 in.
Diameter (Metric) Outer 8 mm
Diameter (Metric) Outer Needle 0.5 mm
Includes Standard plunger
Length (English) Needle 1.96 in.
Product Type Microvolume Syringe
Graduations Major - 25.0 μL, Minor - 5.0 μL
Model 250F
Quantity Ea.
Volume (Metric) 250 μL
Needle Point Style Fixed Bevel
Connection Type Fixed