Learn More
Trajan™ Fixed Needle Syringe for Beckman/Altex, Rheodyne, SSI, Valco Valves
Product Code. 10462721
Click to view available options
Volume (Metric):
25 μL
50 μL
250 μL
500 μL
Unit Size:
Each
Product Code. 10462721
Brand: Trajan™ 004300
Fixed needle replacement for manual syringes. For use with Beckman/Altex, Rheodyne, SSI, and Valco Valves
  • High quality solutions for liquid handling
  • Accurate and reproducible injections
  • Specifically designed for Beckman/Altex, Rheodyne, Valco, SSI valves
Specifications
Material Borosilicate Glass (Barrel)
Needle Gauge 22 G
Diameter (Metric) Inner Needle 0.37 mm
Diameter (Metric) Outer 0.71 mm
Diameter (Metric) Outer Needle 0.028 mm
Product Type Manual Syringe
Volume (Metric) 50 μL
Length (Metric) Needle 51 mm
Needle Point Style LC
For Use With (Equipment) Beckman/Altex, Rheodyne, SSI, Valco Valves