Learn More
Trajan™ PEEKsil™ Tubing
Click to view available options
Diameter (Metric) Inner:
0.05 mm
0.175 mm
Diameter (Metric) Outer:
1.5 mm
1.6 mm
Overall Length (Metric):
50 mm
100 mm
200 mm
500 mm
Unit Size:
Pack of 2
Pack of 5
Description
- Robust - PEEK polymer exterior and fused silica inner tubing
- Smooth wall surface giving excellent flow characteristics
- Compatible with most organic solvents
- Inertness - extremely low absorption characteristics
- Precise ID
Specifications
Specifications
| Type | Tubing |
| Diameter (Metric) Outer | 1.5 mm |
| Diameter (Metric) Inner | 0.05 mm |
| Overall Length (Metric) | 200 mm |